Acid red 1

Supplier: TCI America

Synonyms: Red 2G, C.I. 18050, Amido Naphthol Red G, Sodium 5-acetamido-4-hydroxy-3-(phenyldiazenyl)naphthalene-2,7-disulphonate, 5-Acetamido-4-hydroxy-3-phenylazo-2,7-naphthalene-disulphonic acid disodium salt, Pontacyl Carmine, 5-Acetamido-4-hydroxy-3-phenylazo-2,7-naphthalene-disulfonic acid disodium salt, Azophloxine, Sodium 5-acetamido-4-hydroxy-3-(phenyldiazenyl)naphthalene-2,7-disulfonate

A0585-500G A0585-25G
TCA0585-500GEA 510.34 CAD
TCA0585-500G TCA0585-025G
Acid red 1
Acid red 1

CAS Number: 3734-67-6
MDL Number: MFCD00003954
Molecular Formula: C18H15N3O8S2
Molecular Weight: 509.41
Form: Powder
Color: Red


Other Product Names / Synonyms: Amidonaphthol Red, Azophloxine, Pontacyl Carmine, Red 2G

Formula: C₁₈H₁₅N₃O₈S₂
MW: 509.41 g/mol
MDL Number: MFCD00003954
CAS Number: 3734-67-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8f9113584bacd3bc648d104c2111a4422eb19d87&vid=8f9113584bacd3bc648d104c2111a4422eb19d87&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR