5-Bromo-2,3-pyridinediol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B2780-1G B2780-25G B2780-5G
TCB2780-1GEA 66.04 CAD
TCB2780-1G TCB2780-25G TCB2780-5G
5-Bromo-2,3-pyridinediol ≥98.0% (by GC, titration analysis)
5-Bromo-2,3-pyridinediol

CAS Number: 34206-49-0
Molecular Formula: C5H4BrNO2
Molecular Weight: 190.00
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 249


Other Product Names / Synonyms: 5-Bromo-2,3-dihydroxypyridine

Formula: C₅H₄BrNO₂
Melting Pt: 249 °C
MDL Number: MFCD08276314
CAS Number: 34206-49-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ff5dce9a507fdd0ad1d9541a8ddd971f19ef5705&vid=ff5dce9a507fdd0ad1d9541a8ddd971f19ef5705&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR