6-Bromoindole-3-carboxaldehyde ≥98.0% (by GC, total nitrogen)

Supplier: TCI America

B2778-5G B2778-1G
TCB2778-5GEA 398.16 CAD
TCB2778-5G TCB2778-1G
6-Bromoindole-3-carboxaldehyde ≥98.0% (by GC, total nitrogen)
6-Bromoindole-3-carboxaldehyde

CAS Number: 17826-04-9
Molecular Formula: C9H6BrNO
Molecular Weight: 224.06
Purity/Analysis Method: >98.0% (GC,N)
Form: Crystal
Melting point (°C): 203


Other Product Names / Synonyms: 6-Bromo-3-formylindole

Formula: C₉H₆BrNO
Melting Pt: 203 °C
MDL Number: MFCD00792689
CAS Number: 17826-04-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e686af8d2190b0d3377a77e236ae576f441550fb&vid=e686af8d2190b0d3377a77e236ae576f441550fb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,N)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR