4-sec-Butyl-2,6-di-tert-butylphenol ≥98.0%

Supplier: TCI America

B2774-25G B2774-500G
TCB2774-25GEA 41.71 CAD
TCB2774-25G TCB2774-500G
4-sec-Butyl-2,6-di-tert-butylphenol ≥98.0%
4-sec-Butyl-2,6-di-tert-butylphenol

CAS Number: 17540-75-9
Molecular Formula: C18H30O
Molecular Weight: 262.44
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 141
Flash Point (°C): 113
Freezing point (°C): 24

Formula: C₁₈H₃₀O
Boiling Pt: 141 °C
Flash Pt: 113 °C
MDL Number: MFCD00075575
CAS Number: 17540-75-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db5abb9851de2b41fdda8fe61dc812de0e54a00b&vid=db5abb9851de2b41fdda8fe61dc812de0e54a00b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR