4-Bromoindole-3-carboxyaldehyde ≥96.0% (by GC, total nitrogen)

Supplier: TCI America

B2773-1G B2773-5G
TCB2773-1GEA 81.05 CAD
TCB2773-1G TCB2773-5G
4-Bromoindole-3-carboxyaldehyde ≥96.0% (by GC, total nitrogen)
4-Bromoindole-3-carboxyaldehyde

CAS Number: 98600-34-1
Molecular Formula: C9H6BrNO
Molecular Weight: 224.06
Purity/Analysis Method: >96.0% (GC,N)
Form: Crystal
Melting point (°C): 179


Other Product Names / Synonyms: 4-Bromo-3-formylindole

Formula: C₉H₆BrNO
MW: 224.06 g/mol
Melting Pt: 179 °C
MDL Number: MFCD05864695
CAS Number: 98600-34-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=043966a6a284821784f9828adbad794dcae56b92&vid=043966a6a284821784f9828adbad794dcae56b92&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR