2,4-Bis(ɑ,ɑ-dimethylbenzyl)phenol ≥97.0%

Supplier: TCI America

Synonyms: 2,4-Bis(2-phenylpropan-2-yl)phenol, 2,4-Di-ɑ-cumylphenol

B2772-500G B2772-25G
TCB2772-500GEA 142.2 CAD
TCB2772-500G TCB2772-25G
2,4-Bis(ɑ,ɑ-dimethylbenzyl)phenol ≥97.0%
2,4-Bis(alpha,alpha-dimethylbenzyl)phenol

CAS Number: 2772-45-4
Molecular Formula: C24H26O
Molecular Weight: 330.47
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 221
Melting point (°C): 65


Other Product Names / Synonyms: 2,4-Di-alpha-cumylphenol

Formula: C₂₄H₂₆O
MW: 330.47 g/mol
Boiling Pt: 221 °C
Melting Pt: 65 °C
MDL Number: MFCD00191242
CAS Number: 2772-45-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f548cded257e4266feb19d9ce13fcbcbde6563f7&vid=f548cded257e4266feb19d9ce13fcbcbde6563f7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR