N-Benzylanthranilic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

B2769-25G B2769-5G
TCB2769-25GEA 173.8 CAD
TCB2769-25G TCB2769-5G
N-Benzylanthranilic acid ≥98.0% (by titrimetric analysis)
N-Benzylanthranilic acid

CAS Number: 6622-55-5
MDL Number: MFCD00020247
Molecular Formula: C14H13NO2
Molecular Weight: 227.26
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 171


Other Product Names / Synonyms: 2-(Benzylamino)benzoic Acid

Formula: C₁₄H₁₃NO₂
Melting Pt: 171 °C
MDL Number: MFCD00020247
CAS Number: 6622-55-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c141eab533a37de4c071f1e5d99d304dfbfa6720&vid=c141eab533a37de4c071f1e5d99d304dfbfa6720&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR