4,6-Bis(difluoromethoxy)-2-(methylthio)pyrimidine ≥98.0%

Supplier: TCI America

B2755-25G B2755-5G
TCB2755-25GEA 247.9 CAD
TCB2755-25G TCB2755-5G
4,6-Bis(difluoromethoxy)-2-(methylthio)pyrimidine ≥98.0%
4,6-Bis(difluoromethoxy)-2-(methylthio)pyrimidine

CAS Number: 100478-25-9
Molecular Formula: C7H6F4N2O2S
Molecular Weight: 258.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 48
Flash Point (°C): 133

Formula: C₇H₆F₄N₂O₂S
MW: 258.19 g/mol
Melting Pt: 48 °C
Flash Pt: 133 °C
CAS Number: 100478-25-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ad10707c81772aa0f3135dc014d697141fad780&vid=6ad10707c81772aa0f3135dc014d697141fad780&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR