Butylsuccinic anhydride ≥98.0%

Supplier: TCI America

B2742-5G B2742-25G
TCB2742-5GEA 285.98 CAD
TCB2742-5G TCB2742-25G
Butylsuccinic anhydride ≥98.0%
Butylsuccinic anhydride

CAS Number: 2035-76-9
Molecular Formula: C8H12O3
Molecular Weight: 156.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 46
Storage Temperature: 0-10°C

Formula: C₈H₁₂O₃
Melting Pt: 46 °C
CAS Number: 2035-76-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b7f013affa13e8db47d71dc45f1c6fdb1a55a748&vid=b7f013affa13e8db47d71dc45f1c6fdb1a55a748&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR