4,4'-Bis(2-methoxystyryl)biphenyl ≥90.0%
Supplier: TCI America
Synonyms:
4,4-Bis(2-methoxystyryl)biphenyl, 4,4'-Bis[2-(2-methoxyphenyl)vinyl]biphenyl
B2730-500G
B2730-25G
TCB2730-500GEA
703.1
CAD
TCB2730-500G
TCB2730-25G
4,4'-Bis(2-methoxystyryl)biphenyl ≥90.0%
4,4'-Bis(2-methoxystyryl)biphenyl
CAS Number: 40470-68-6
Molecular Formula: C30H26O2
Molecular Weight: 418.54
Purity/Analysis Method: >90.0% (HPLC)
Form: Crystal
Melting point (°C): 218
Formula:
C₃₀H₂₆O₂ MW: 418.54 g/mol Melting Pt: 218 °C |
MDL Number:
MFCD23160777 CAS Number: 40470-68-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...