1,4-Bis(2-benzoxazolyl)naphthalene ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

B2728-25G B2728-500G
TCB2728-25GEA 84.37 CAD
TCB2728-25G TCB2728-500G
1,4-Bis(2-benzoxazolyl)naphthalene ≥98.0% (by HPLC, total nitrogen)
1,4-Bis(2-benzoxazolyl)naphthalene

CAS Number: 5089-22-5
Molecular Formula: C24H14N2O2
Molecular Weight: 362.39
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 212

Formula: C₂₄H₁₄N₂O₂
Melting Pt: 212 °C
MDL Number: MFCD00357168
CAS Number: 5089-22-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9d3176571ae7a4fad33ef1a5b1027a38168df56&vid=e9d3176571ae7a4fad33ef1a5b1027a38168df56&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR