Bis(2-butoxyethyl)adipate ≥97.0%
Supplier: TCI America
Synonyms:
Di(2-butoxyethyl) Adipate, Adipic Acid Bis(2-butoxyethyl) Ester, Adipic Acid Di(2-butoxyethyl) Ester
CAS Number: 141-18-4
Molecular Formula: C18H34O6
Molecular Weight: 346.46
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 217
Melting point (°C): -34
Flash Point (°C): 199
Specific Gravity (20/20): 1.00
Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: Adipic Acid Bis(2-butoxyethyl) Ester, Adipic Acid Di(2-butoxyethyl) Ester, Di(2-butoxyethyl) Adipate
Formula:
C₁₈H₃₄O₆ MW: 346.46 g/mol Boiling Pt: 217 °C Melting Pt: –34 °C Density: 1.00 (20 °C) Flash Pt: 199 °C |
CAS Number:
141-18-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...