4,4'-Bis(9H-carbazol-9-yl)biphenyl ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
Synonyms:
CBP, 4,4'-Di(9H-carbazol-9-yl)-1,1'-biphenyl, 4,4-Di(9H-carbazol-9-yl)-1,1-biphenyl
CAS Number: 58328-31-7
MDL Number: MFCD00093417
Molecular Formula: C36H24N2
Molecular Weight: 484.60
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 283
Lambda max.: 319 nm (CH2Cl2)
Product Categories: Materials Chemistry, Organic Light-Emitting Diode (OLED) Materials
Other Product Names / Synonyms: CBP, 4,4'-Di(9H-carbazol-9-yl)-1,1'-biphenyl
Formula:
C₃₆H₂₄N₂ MW: 484.60 g/mol Melting Pt: 283 °C |
MDL Number:
MFCD00093417 CAS Number: 58328-31-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...