Benzyl-2-chloroethyl ether ≥98.0%
Supplier: TCI America
Synonyms:
2-(Benzyloxy)ethyl Chloride, ((2-Chloroethoxy)methyl)benzene, (2-Chloroethoxymethyl)benzene
B2712-5G
B2712-25G
TCB2712-5GEA
188.02
CAD
TCB2712-5G
TCB2712-25G
Benzyl-2-chloroethyl ether ≥98.0%
Benzyl-2-chloroethyl ether
CAS Number: 17229-17-3
Molecular Formula: C9H11ClO
Molecular Weight: 170.64
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.11
Other Product Names / Synonyms: 2-(Benzyloxy)ethyl Chloride, (2-Chloroethoxymethyl)benzene
Formula:
C₉H₁₁ClO MW: 170.64 g/mol Density: 1.11 (20 °C) |
MDL Number:
MFCD00018964 CAS Number: 17229-17-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...