N-(4-Bromophenyl)phthalimide ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
B2707-25G
B2707-5G
TCB2707-25GEA
550.47
CAD
TCB2707-25G
TCB2707-5G
N-(4-Bromophenyl)phthalimide ≥98.0% (by HPLC, total nitrogen)
N-(4-Bromophenyl)phthalimide
CAS Number: 40101-31-3
Molecular Formula: C14H8BrNO2
Molecular Weight: 302.13
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 206
Formula:
C₁₄H₈BrNO₂ Melting Pt: 206 °C |
MDL Number:
MFCD00047301 CAS Number: 40101-31-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...