2-Bromo-9,10-anthraquinone ≥96.0%
Supplier: TCI America
Synonyms:
2-Bromoanthracene-9,10-dione, 2-Bromo-9,10-anthracenedione
B2689-1G
B2689-25G
B2689-5G
TCB2689-1GEA
71.1
CAD
TCB2689-1G
TCB2689-25G
TCB2689-5G
2-Bromo-9,10-anthraquinone ≥96.0%
2-Bromo-9,10-anthraquinone
CAS Number: 572-83-8
Molecular Formula: C14H7BrO2
Molecular Weight: 287.11
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Melting point (°C): 208
Formula:
C₁₄H₇BrO₂ MW: 287.11 g/mol Melting Pt: 208 °C |
MDL Number:
MFCD00474541 CAS Number: 572-83-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...