1-Bromo-9,10-anthraquinone ≥95.0%
Supplier: TCI America
B2688-1G
B2688-5G
TCB2688-1GEA
246.48
CAD
TCB2688-1G
TCB2688-5G
1-Bromo-9,10-anthraquinone ≥95.0%
1-Bromo-9,10-anthraquinone
CAS Number: 632-83-7
Molecular Formula: C14H7BrO2
Molecular Weight: 287.11
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 191
Formula:
C₁₄H₇BrO₂ Melting Pt: 191 °C |
MDL Number:
MFCD00451689 CAS Number: 632-83-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...