Bis(4-bromophenyl)amine ≥98.0%

Supplier: TCI America

Synonyms: 4,4'-Dibromodiphenylamine

B2684-5G B2684-1G
TCB2684-5GEA 82.16 CAD
TCB2684-5G TCB2684-1G
Bis(4-bromophenyl)amine ≥98.0%
Bis(4-bromophenyl)amine

CAS Number: 16292-17-4
Molecular Formula: C12H9Br2N
Molecular Weight: 327.02
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 106


Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors
Other Product Names / Synonyms: 4,4'-Dibromodiphenylamine

Formula: C₁₂H₉Br₂N
MW: 327.02 g/mol
Melting Pt: 106 °C
MDL Number: MFCD00225488
CAS Number: 16292-17-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=65ce8a211a6f7c6c57c29a53bc1451973e57b92f&vid=65ce8a211a6f7c6c57c29a53bc1451973e57b92f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR