1-Butyl-3-methylimidazoliumtetrachloroferrate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

B2672-25G B2672-5G
TCB2672-25GEA 883.22 CAD
TCB2672-25G TCB2672-5G
1-Butyl-3-methylimidazoliumtetrachloroferrate ≥98.0% (by titrimetric analysis)
1-Butyl-3-methylimidazoliumtetrachloroferrate

CAS Number: 359845-21-9
Molecular Formula: C8H15Cl4FeN2
Molecular Weight: 336.87
Purity/Analysis Method: >98.0% (T)
Form: Liquid

Formula: C₈H₁₅Cl₄FeN₂
MDL Number: MFCD08276298
CAS Number: 359845-21-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aa6f031c7c317a6c7fdf207c7eee19b163dfded8&vid=aa6f031c7c317a6c7fdf207c7eee19b163dfded8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR