1,3-Bis[2-(4-hydroxyphenyl)-2-propyl]benzene ≥98.0%

Supplier: TCI America

Synonyms: 4,4'-(1,3-Phenylenediisopropylidene)bisphenol, Bisphenol M

B2664-25G B2664-250G
TCB2664-25GEA 104.64 CAD
TCB2664-25G TCB2664-250G
1,3-Bis[2-(4-hydroxyphenyl)-2-propyl]benzene ≥98.0%
1,3-Bis[2-(4-hydroxyphenyl)-2-propyl]benzene

CAS Number: 13595-25-0
Molecular Formula: C24H26O2
Molecular Weight: 346.47
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 138


Product Categories: Materials Chemistry, Monomers, Pressure & Heat Sensitive Recording Materials
Other Product Names / Synonyms: Bisphenol M, 4,4'-(1,3-Phenylenediisopropylidene)bisphenol

Formula: C₂₄H₂₆O₂
MW: 346.47 g/mol
Melting Pt: 138 °C
CAS Number: 13595-25-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=818f639ce69e56d85ce0796b1f5de975defd2185&vid=818f639ce69e56d85ce0796b1f5de975defd2185&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR