3,5-Bis(tert-butylthio)-1-chlorobenzene ≥98.0%

Supplier: TCI America

Synonyms: 1-Chloro-3,5-bis(tert-butylthio)benzene

B2659-5G B2659-1G
TCB2659-5GEA 64.78 CAD
TCB2659-5G TCB2659-1G
3,5-Bis(tert-butylthio)-1-chlorobenzene ≥98.0%
3,5-Bis(tert-butylthio)-1-chlorobenzene

CAS Number: 260968-02-3
Molecular Formula: C14H21ClS2
Molecular Weight: 288.89
Purity/Analysis Method: >98.0% (GC)
Form: Crystal


Other Product Names / Synonyms: 1-Chloro-3,5-bis(tert-butylthio)benzene

Formula: C₁₄H₂₁ClS₂
CAS Number: 260968-02-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=145b7c23ecd2128c5972cf73cbbc56067e054622&vid=145b7c23ecd2128c5972cf73cbbc56067e054622&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR