4-Bromo-N,N-diphenylaniline ≥97.0%
Supplier: TCI America
B2633-25G
B2633-5G
TCB2633-25GEA
150.1
CAD
TCB2633-25G
TCB2633-5G
4-Bromo-N,N-diphenylaniline ≥97.0%
4-Bromo-N,N-diphenylaniline
CAS Number: 36809-26-4
Molecular Formula: C18H14BrN
Molecular Weight: 324.22
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 185
Melting point (°C): 115
Formula:
C₁₈H₁₄BrN Boiling Pt: 185 °C Melting Pt: 115 °C |
MDL Number:
MFCD01851266 CAS Number: 36809-26-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...