cis-4-tert-Butylcyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B2631-25G B2631-5G
TCB2631-25GEA 995.4 CAD
TCB2631-25G TCB2631-5G
cis-4-tert-Butylcyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)
cis-4-tert-Butylcyclohexanecarboxylic acid

CAS Number: 943-28-2
Molecular Formula: C11H20O2
Molecular Weight: 184.28
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 117

Formula: C₁₁H₂₀O₂
Melting Pt: 117 °C
CAS Number: 943-28-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e168c14ac45de3709a3c50251d27380db33ac310&vid=e168c14ac45de3709a3c50251d27380db33ac310&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR