1,2-Bis(2,4-dimethyl-5-phenyl-3-thienyl)-3,3,4,4,5,5-hexafluoro-1-cyclopentene ≥98.0%

Supplier: TCI America

B2629-100MG B2629-1G
TCB2629-100MGEA 201.61 CAD
TCB2629-100MG TCB2629-1G
1,2-Bis(2,4-dimethyl-5-phenyl-3-thienyl)-3,3,4,4,5,5-hexafluoro-1-cyclopentene ≥98.0%
1,2-Bis(2,4-dimethyl-5-phenyl-3-thienyl)-3,3,4,4,5,5-hexafluoro-1-cyclopentene

CAS Number: 172612-67-8
Molecular Formula: C29H22F6S2
Molecular Weight: 548.61
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 132

Formula: C₂₉H₂₂F₆S₂
MW: 548.61 g/mol
Melting Pt: 132 °C
CAS Number: 172612-67-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=502134066592b313c3ad1430aae9bbdd8a78dbea&vid=502134066592b313c3ad1430aae9bbdd8a78dbea&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR