Bis(2,2,2-trifluoroethyl)phosphite ≥94.0%
Supplier: TCI America
Synonyms:
Phosphorous Acid Bis(2,2,2-trifluoroethyl) Ester, Bis(2,2,2-trifluoroethyl) Phosphonate, Phosphonic Acid Bis(2,2,2-trifluoroethyl) Ester
CAS Number: 92466-70-1
Molecular Formula: C4H5F6O3P
Molecular Weight: 246.05
Purity/Analysis Method: >94.0% (GC)
Form: Clear Liquid
Boiling point (°C): 44
Flash Point (°C): 76
Specific Gravity (20/20): 1.56
Other Product Names / Synonyms: Bis(2,2,2-trifluoroethyl) Phosphonate, Phosphonic Acid Bis(2,2,2-trifluoroethyl) Ester, Phosphorous Acid Bis(2,2,2-trifluoroethyl) Ester
Formula:
C₄H₅F₆O₃P MW: 246.05 g/mol Boiling Pt: 44 °C Density: 1.56 (20 °C) Flash Pt: 76 °C |
MDL Number:
MFCD00063314 CAS Number: 92466-70-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...