2-Bromo-3-formylpyridine ≥98.0%
Supplier: TCI America
Synonyms:
2-Bromopyridine-3-carboxaldehyde, 2-Bromonicotinaldehyde, 2-Bromo-3-pyridinecarboxaldehyde
B2625-1G
B2625-5G
TCB2625-1GEA
52.14
CAD
TCB2625-1G
TCB2625-5G
2-Bromo-3-formylpyridine ≥98.0%
2-Bromo-3-formylpyridine
CAS Number: 128071-75-0
Molecular Formula: C6H4BrNO
Molecular Weight: 186.01
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 100
Melting point (°C): 73
Other Product Names / Synonyms: 2-Bromopyridine-3-carboxaldehyde
Formula:
C₆H₄BrNO MW: 186.01 g/mol Boiling Pt: 100 °C Melting Pt: 73 °C |
MDL Number:
MFCD04966945 CAS Number: 128071-75-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...