N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,1,2,2-tetramethylethylenediamine ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
B2619-1G
B2619-5G
TCB2619-1GEA
145.36
CAD
TCB2619-1G
TCB2619-5G
N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,1,2,2-tetramethylethylenediamine ≥98.0% (by HPLC, total nitrogen)
N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,1,2,2-tetramethylethylenediamine
CAS Number: 351498-10-7
MDL Number: MFCD06797077
Molecular Formula: C36H56N2O2
Molecular Weight: 548.86
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Formula:
C₃₆H₅₆N₂O₂ |
MDL Number:
MFCD06797077 CAS Number: 351498-10-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...