2-Bromo-6-benzyloxypyridine ≥97.0%

Supplier: TCI America

Synonyms: 2-(Benzyloxy)-6-bromopyridine

B2576-25G B2576-5G
TCB2576-25GEA 699.15 CAD
TCB2576-25G TCB2576-5G
2-Bromo-6-benzyloxypyridine ≥97.0%
2-Bromo-6-benzyloxypyridine

CAS Number: 117068-71-0
Molecular Formula: C12H10BrNO
Molecular Weight: 264.12
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Freezing point (°C): 36
Storage Temperature: 0-10°C

Formula: C₁₂H₁₀BrNO
MW: 264.12 g/mol
MDL Number: MFCD00234782
CAS Number: 117068-71-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9108e9761fcc4b4c77f490bfe489a1389282eaa9&vid=9108e9761fcc4b4c77f490bfe489a1389282eaa9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR