Bis(vinylsulfonyl)methane ≥96.0%

Supplier: TCI America

B2550-25G B2550-5G
TCB2550-25GEA 331.8 CAD
TCB2550-25G TCB2550-5G
Bis(vinylsulfonyl)methane ≥96.0%
Bis(vinylsulfonyl)methane

CAS Number: 3278-22-6
MDL Number: MFCD03265421
Molecular Formula: C5H8O4S2
Molecular Weight: 196.24
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Melting point (°C): 62

Formula: C₅H₈O₄S₂
MW: 196.24 g/mol
Melting Pt: 62 °C
MDL Number: MFCD03265421
CAS Number: 3278-22-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4efa5e1034f2ef5de260299fc956c2b854403c97&vid=4efa5e1034f2ef5de260299fc956c2b854403c97&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR