1,4-Bis(trifluoromethyl)-2-nitrobenzene ≥98.0%

Supplier: TCI America

Synonyms: 2-Nitro-1,4-bis(trifluoromethyl)benzene, 1-Nitro-2,5-bis(trifluoromethyl)benzene

B2546-25G B2546-5G
TCB2546-25GEA 128.93 CAD
TCB2546-25G TCB2546-5G
1,4-Bis(trifluoromethyl)-2-nitrobenzene ≥98.0%
1,4-Bis(trifluoromethyl)-2-nitrobenzene

CAS Number: 320-88-7
MDL Number: MFCD00236674
Molecular Formula: C8H3F6NO2
Molecular Weight: 259.11
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 90
Specific Gravity (20/20): 1.57

Formula: C₈H₃F₆NO₂
MW: 259.11 g/mol
Boiling Pt: 90 °C
Density: 1.57 (20 °C)
MDL Number: MFCD00236674
CAS Number: 320-88-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d19b4767db0e88c4f6cf116cec911ddae9fe5c7c&vid=d19b4767db0e88c4f6cf116cec911ddae9fe5c7c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR