Bis(N,N-dimethylacetamide) hydrogen dibromobromate ≥97.0% (by titrimetric analysis)

Supplier: TCI America

B2509-250G B2509-25G
TCB2509-250GEA 285.98 CAD
TCB2509-250G TCB2509-25G
Bis(N,N-dimethylacetamide) hydrogen dibromobromate ≥97.0% (by titrimetric analysis)
Bis(N,N-dimethylacetamide) hydrogen dibromobromate

CAS Number: 75381-80-5
Molecular Formula: C8H19Br3N2O2
Molecular Weight: 414.96
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 92


Other Product Names / Synonyms: Bis(N,N-dimethylacetamide) Hydrogen Tribromide

Formula: C₈H₁₉Br₃N₂O₂
Melting Pt: 92 °C
CAS Number: 75381-80-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=89169f1166548523cffa82912bc40ef202a3b29f&vid=89169f1166548523cffa82912bc40ef202a3b29f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR