4-Benzylthiomorpholine-1,1-dioxide ≥98.0%

Supplier: TCI America

B2434-5G B2434-25G
TCB2434-5GEA 87.53 CAD
TCB2434-5G TCB2434-25G
4-Benzylthiomorpholine-1,1-dioxide ≥98.0%
4-Benzylthiomorpholine-1,1-dioxide

CAS Number: 26475-66-1
Molecular Formula: C11H15NO2S
Molecular Weight: 225.31
Purity/Analysis Method: >98.0% (GC)
Form: Crystal

Formula: C₁₁H₁₅NO₂S
CAS Number: 26475-66-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=052626b7cf364ee224ca362e20feb472d617b7d7&vid=052626b7cf364ee224ca362e20feb472d617b7d7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR