Bis(4-biphenylyl)amine ≥98.0%
Supplier: TCI America
B2429-1G
B2429-5G
TCB2429-1GEA
47.87
CAD
TCB2429-1G
TCB2429-5G
Bis(4-biphenylyl)amine ≥98.0%
Bis(4-biphenylyl)amine
CAS Number: 102113-98-4
Molecular Formula: C24H19N
Molecular Weight: 321.42
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 209
Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors
Other Product Names / Synonyms: 4,4'-Iminobis(biphenyl), Di([1,1'-biphenyl]-4-yl)amine
Formula:
C₂₄H₁₉N Melting Pt: 209 °C |
MDL Number:
MFCD08276279 CAS Number: 102113-98-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...