4-Bromoisatin ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 4-Bromoindoline-2,3-dione

B2423-25G B2423-5G
TCB2423-25GEA 184.86 CAD
TCB2423-25G TCB2423-5G
4-Bromoisatin ≥98.0% (by GC, titration analysis)
4-Bromoisatin

CAS Number: 20780-72-7
Molecular Formula: C8H4BrNO2
Molecular Weight: 226.03
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 271


Product Categories: Synthetic Organic Chemistry, Isatins (Multi-component Reaction)
Other Product Names / Synonyms: 4-Bromoindole-2,3-dione, 4-Bromo-2,3-indolinedione

Formula: C₈H₄BrNO₂
MW: 226.0269 g/mol
Melting Pt: 271 °C
MDL Number: MFCD03618555
CAS Number: 20780-72-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=73021bac73fffd6b4de887d9383bfb04a8f8e7f1&vid=73021bac73fffd6b4de887d9383bfb04a8f8e7f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR