tert-Butyl-N-(2-hydroxypropyl)carbamate ≥98.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: N-(2-Hydroxypropyl)carbamic Acid tert-Butyl Ester, 1-(tert-Butoxycarbonylamino)-2-propanol, 1-(Boc-amino)-2-propanol, tert-Butyl (2-hydroxypropyl)carbamate

B2418-5G B2418-25G
TCB2418-5GEA 170.64 CAD
TCB2418-5G TCB2418-25G
tert-Butyl-N-(2-hydroxypropyl)carbamate ≥98.0% (by total nitrogen basis)
tert-Butyl-N-(2-hydroxypropyl)carbamate

CAS Number: 95656-86-3
Molecular Formula: C8H17NO3
Molecular Weight: 175.23
Purity/Analysis Method: >98.0% (N)
Form: Clear Liquid
Specific Gravity (20/20): 1.02


Other Product Names / Synonyms: 1-(Boc-amino)-2-propanol, 1-(tert-Butoxycarbonylamino)-2-propanol, N-(2-Hydroxypropyl)carbamic Acid tert-Butyl Ester

Formula: C₈H₁₇NO₃
MW: 175.23 g/mol
Density: 1.02 (20 °C)
MDL Number: MFCD02093569
CAS Number: 95656-86-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=28c433ddb9955cf1ec6eb239d7f332b33ba3d1dd&vid=28c433ddb9955cf1ec6eb239d7f332b33ba3d1dd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR