4-Bromo-trans-stilbene ≥98.0%
Supplier: TCI America
B2416-1G
B2416-5G
TCB2416-1GEA
139.04
CAD
TCB2416-1G
TCB2416-5G
4-Bromo-trans-stilbene ≥98.0%
4-Bromo-trans-stilbene
CAS Number: 4714-24-3
MDL Number: MFCD00155020
Molecular Formula: C14H11Br
Molecular Weight: 259.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 140
Formula:
C₁₄H₁₁Br Melting Pt: 140 °C |
MDL Number:
MFCD00155020 CAS Number: 13041-70-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...