12-Benzylamino-1-dodecanol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

B2403-1G B2403-5G
TCB2403-1GEA 221.52 CAD
TCB2403-1G TCB2403-5G
12-Benzylamino-1-dodecanol ≥98.0% (by GC, titration analysis)
12-Benzylamino-1-dodecanol

MDL Number: MFCD06797075
Molecular Formula: C19H33NO
Molecular Weight: 291.48
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal


Other Product Names / Synonyms: N-(12-Hydroxydodecyl)benzylamine

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f4a81c2eede85fbc02a60673eaa08f139952b55&vid=1ba7de3668b6a79b8572139c17eac09a7600b7a9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR