9,9-Bis(4-hydroxy-3-methylphenyl)fluorene ≥98.0%
Supplier: TCI America
B2396-250G
B2396-25G
TCB2396-250GEA
525.67
CAD
TCB2396-250G
TCB2396-25G
9,9-Bis(4-hydroxy-3-methylphenyl)fluorene ≥98.0%
9,9-Bis(4-hydroxy-3-methylphenyl)fluorene
CAS Number: 88938-12-9
MDL Number: MFCD04038046
Molecular Formula: C27H22O2
Molecular Weight: 378.47
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 219
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 4,4'-(9-Fluorenylidene)di(o-cresol)
Formula:
C₂₇H₂₂O₂ Melting Pt: 219 °C |
MDL Number:
MFCD04038046 CAS Number: 88938-12-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...