1-Boc-4-(hydroxymethyl)piperidine ≥98.0%

Supplier: TCI America

Synonyms: tert-butyl 4-(hydroxymethyl)piperidine-1-carboxylate, 1-Boc-4-piperidinemethanol, N-BOC-4-Piperidinemethanol

B2364-5G B2364-1G B2364-25G
TCB2364-5GEA 52.14 CAD
TCB2364-5G TCB2364-1G TCB2364-25G
1-Boc-4-(hydroxymethyl)piperidine ≥98.0%
1-Boc-4-(hydroxymethyl)piperidine

CAS Number: 123855-51-6
MDL Number: MFCD02094488
Molecular Formula: C11H21NO3
Molecular Weight: 215.29
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 79


Other Product Names / Synonyms: 1-Boc-4-(hydroxymethyl)piperidine, 1-Boc-4-piperidinemethanol, 1-(tert-Butoxycarbonyl)-4-(hydroxymethyl)piperidine

Formula: C₁₁H₂₁NO₃
MW: 215.29 g/mol
Melting Pt: 79 °C
MDL Number: MFCD02094488
CAS Number: 123855-51-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=04916a4bcfc763b92db30a1adcdbc983e80c7621&vid=04916a4bcfc763b92db30a1adcdbc983e80c7621&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR