Bis(2,4,6-trimethylpyridine)iodonium hexafluorophosphate ≥98.0%
Supplier: TCI America
B2359-5G
B2359-1G
TCB2359-5GEA
342.86
CAD
TCB2359-5G
TCB2359-1G
Bis(2,4,6-trimethylpyridine)iodonium hexafluorophosphate ≥98.0%
Bis(2,4,6-trimethylpyridine)iodonium hexafluorophosphate
CAS Number: 113119-46-3
Molecular Formula: C16H22F6IN2P
Molecular Weight: 514.23
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 126
Storage Temperature: 0-10°C
Formula:
C₁₆H₂₂F₆IN₂P Melting Pt: 126 °C |
CAS Number:
113119-46-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...