Bis(2,4,6-trimethylpyridine)bromonium hexafluorophosphate ≥98.0% (by HPLC)
Supplier: TCI America
B2358-1G
B2358-5G
TCB2358-1GEA
145.36
CAD
TCB2358-1G
TCB2358-5G
Bis(2,4,6-trimethylpyridine)bromonium hexafluorophosphate ≥98.0% (by HPLC)
Bis(2,4,6-trimethylpyridine)bromonium hexafluorophosphate
CAS Number: 188944-77-6
Molecular Formula: C16H22BrF6N2P
Molecular Weight: 467.23
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Storage Temperature: 0-10°C
Formula:
C₁₆H₂₂BrF₆N₂P |
CAS Number:
188944-77-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...