3-Bromo-3,3-difluoropropene ≥95.0%

Supplier: TCI America

Synonyms: 3,3-Difluoro-3-bromopropene

B2333-1G B2333-5G
TCB2333-1GEA 328.64 CAD
TCB2333-1G TCB2333-5G
3-Bromo-3,3-difluoropropene ≥95.0%
3-Bromo-3,3-difluoropropene

CAS Number: 420-90-6
MDL Number: MFCD00042185
Molecular Formula: C3H3BrF2
Molecular Weight: 156.96
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 42
Specific Gravity (20/20): 1.55
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorinated Building Blocks
Other Product Names / Synonyms: 3,3-Difluoro-3-bromopropene

Formula: C₃H₃BrF₂
MW: 156.96 g/mol
Boiling Pt: 42 °C
Density: 1.55 (20 °C)
MDL Number: MFCD00042185
CAS Number: 420-90-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=040be4e09d42abdd421731796814acb66b7a410b&vid=040be4e09d42abdd421731796814acb66b7a410b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR