Ethyl-9-bromononanoate ≥95.0%

Supplier: TCI America

B2324-1G B2324-5G
TCB2324-1GEA 297.04 CAD
TCB2324-1G TCB2324-5G
Ethyl-9-bromononanoate ≥95.0%
Ethyl-9-bromononanoate

CAS Number: 28598-81-4
MDL Number: MFCD06797065
Molecular Formula: C11H21BrO2
Molecular Weight: 265.19
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 180
Specific Gravity (20/20): 1.17


Other Product Names / Synonyms: 9-Bromononanoic Acid Ethyl Ester

Formula: C₁₁H₂₁BrO₂
Boiling Pt: 180 °C
Density: 1.17 (20 °C)
MDL Number: MFCD06797065
CAS Number: 28598-81-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9dbb3b90de863cac5e62b0f45d606c113a806b95&vid=9dbb3b90de863cac5e62b0f45d606c113a806b95&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR