(+/-)-1,2-Bis(2,4,6-trimethylphenyl)ethylenediamine dihydrochloride ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
B2318-5G
B2318-1G
TCB2318-5GEA
1757.12
CAD
TCB2318-5G
TCB2318-1G
(+/-)-1,2-Bis(2,4,6-trimethylphenyl)ethylenediamine dihydrochloride ≥98.0% (by HPLC, total nitrogen)
(+/-)-1,2-Bis(2,4,6-trimethylphenyl)ethylenediamine dihydrochloride
Molecular Weight: 369.37
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Molecular Formula: C20H28N2
Other Product Names / Synonyms: (+/-)-1,2-Dimesitylethylenediamine Dihydrochloride, (+/-)-1,2-Diamino-1,2-dimesitylethane Dihydrochloride
Formula:
C₂₀H₂₈N₂.2HCl |
CAS Number:
1820706-16-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...