4-tert-Butylsulfonylcalix[4]arene ≥98.0% (by HPLC)
Supplier: TCI America
B2312-1G
B2312-5G
TCB2312-1GEA
377.62
CAD
TCB2312-1G
TCB2312-5G
4-tert-Butylsulfonylcalix[4]arene ≥98.0% (by HPLC)
4-tert-Butylsulfonylcalix[4]arene
CAS Number: 204190-49-8
MDL Number: MFCD06797060
Molecular Formula: C40H48O12S4
Molecular Weight: 849.05
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Product Categories: Materials Chemistry, Macrocycles for Host-Guest Chemistry
Other Product Names / Synonyms: Tetra-tert-butyl(tetrahydroxy)tetrasulfonylcalix[4]arene
Formula:
C₄₀H₄₈O₁₂S₄ |
MDL Number:
MFCD06797060 CAS Number: 204190-49-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...