4-tert-Butyl-1-(ethoxycarbonylmethoxy)thiacalix[4]arene ≥94.0% (by HPLC)

Supplier: TCI America

Synonyms: 4-tert-Butyl-1-(Ethoxycarbonylmethoxy)thiacalix[4]arene, Tetra-tert-butyl[tetrakis(ethoxycarbonylmethoxy)]tetrathiacalix[4]arene

B2310-1G B2310-5G
TCB2310-1GEA 142.2 CAD
TCB2310-1G TCB2310-5G
4-tert-Butyl-1-(ethoxycarbonylmethoxy)thiacalix[4]arene ≥94.0% (by HPLC)
4-tert-Butyl-1-(ethoxycarbonylmethoxy)thiacalix[4]arene

CAS Number: 210706-03-9
MDL Number: MFCD06797059
Molecular Formula: C56H72O12S4
Molecular Weight: 1065.42
Purity/Analysis Method: >94.0% (HPLC)
Form: Crystal


Product Categories: Materials Chemistry, Macrocycles for Host-Guest Chemistry
Other Product Names / Synonyms: Tetra-tert-butyl[tetrakis(ethoxycarbonylmethoxy)]tetrathiacalix[4]arene

Formula: C₅₆H₇₂O₁₂S₄
MW: 1065.42 g/mol
MDL Number: MFCD06797059
CAS Number: 210706-03-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d73a37d91a603b9fa31d673c12dc780fd25f2f1&vid=3d73a37d91a603b9fa31d673c12dc780fd25f2f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 94.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR