4,4'-Azoxyanisole ≥98.0%

Supplier: TCI America

A0554-25G A0554-5G
TCA0554-025GEA 272.34 CAD
TCA0554-025G TCA0554-5G
4,4'-Azoxyanisole ≥98.0%
4,4'-Azoxyanisole

CAS Number: 1562-94-3
MDL Number: MFCD00008400
Molecular Formula: C14H14N2O3
Molecular Weight: 258.28
Purity/Analysis Method: >98.0% (GC)
Form: Crystal


Product Categories: Materials Chemistry, Liquid Crystal Materials (Optically Functional Materials)
Other Product Names / Synonyms: 4,4'-Dimethoxyazoxybenzene

Formula: C₁₄H₁₄N₂O₃
MDL Number: MFCD00008400
CAS Number: 1562-94-3
EINECS: 216-348-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=079c9059d4f22004a3f91d3e2c6413341a0802cf&vid=079c9059d4f22004a3f91d3e2c6413341a0802cf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR