4-tert-Butylthiacalix[4]arene ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
Tetra-tert-butyl(tetrahydroxy)tetrathiacalix[4]arene
B2296-5G
B2296-1G
TCB2296-5GEA
271.76
CAD
TCB2296-5G
TCB2296-1G
4-tert-Butylthiacalix[4]arene ≥98.0% (by HPLC)
4-tert-Butylthiacalix[4]arene
CAS Number: 182496-55-5
MDL Number: MFCD04038412
Molecular Formula: C40H48O4S4
Molecular Weight: 721.06
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 323
Product Categories: Materials Chemistry, Macrocycles for Host-Guest Chemistry
Other Product Names / Synonyms: Tetra-tert-butyl(tetrahydroxy)tetrathiacalix[4]arene
Formula:
C₄₀H₄₈O₄S₄ MW: 721.06 g/mol Melting Pt: 323 °C |
MDL Number:
MFCD04038412 CAS Number: 182496-55-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...