tert-Butyl (2-bromoethyl)carbamate ≥98.0%
Supplier: TCI America
B2289-1G
B2289-5G
B2289-25G
TCB2289-1GEA
104.53
CAD
TCB2289-1G
TCB2289-5G
TCB2289-25G
tert-Butyl (2-bromoethyl)carbamate ≥98.0%
tert-Butyl (2-bromoethyl)carbamate
CAS Number: 39684-80-5
MDL Number: MFCD02683428
Molecular Formula: C7H14BrNO2
Molecular Weight: 224.10
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 33
Storage Temperature: <0°C
Other Product Names / Synonyms: tert-Butyl N-(2-Bromoethyl)carbamate, N-(2-Bromoethyl)carbamic Acid tert-Butyl Ester, 2-(Boc-amino)ethyl Bromide
Formula:
C₇H₁₄BrNO₂ Melting Pt: 33 °C |
MDL Number:
MFCD02683428 CAS Number: 39684-80-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...