1,2-Bis[2-methylbenzo[b]thiophen-3-yl]-3,3,4,4,5,5-hexafluoro-1-cyclopentene ≥97.0%

Supplier: TCI America

B2287-100MG B2287-1G
TCB2287-100MGEA 109.02 CAD
TCB2287-100MG TCB2287-1G
1,2-Bis[2-methylbenzo[b]thiophen-3-yl]-3,3,4,4,5,5-hexafluoro-1-cyclopentene ≥97.0%

CAS Number: 137814-07-4
MDL Number: MFCD06656279
Molecular Formula: C23H14F6S2
Molecular Weight: 468.48
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 160

Formula: C₂₃H₁₄F₆S₂
Melting Pt: 160 °C
MDL Number: MFCD06656279
CAS Number: 137814-07-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1a903fb9e006e684937102f165b8423f22667fb1&vid=1a903fb9e006e684937102f165b8423f22667fb1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR